Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213897.1 | NP_999062.1 | 8 - 9 | 982 | 150 | |
GenBank mRNA | D89478.1 | - | 8 - 9 | 982 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |