Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213947.1 | NP_999112.1 | 2 - 3 | 247 | 130 | |
GenBank mRNA | AJ506744.1 | - | 2 - 3 | 247 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |