Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348975.2 | NP_001335904.1 | 9 - 10 | 1609 | 87 | |
GenBank mRNA | AK346291.1 | - | 9 - 10 | 1609 | 87 | |
AK394181.1 | - | 8 - 9 | 1091 | 87 | ||
AY238476.2 | - | 9 - 10 | 1581 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |