Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP91-PHOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214043.2 | NP_999208.1 | 6 - 7 | 899 | 121 | |
GenBank mRNA | AK230821.1 | - | 9 - 10 | 1230 | 121 | |
AK231643.1 | - | 9 - 10 | 1231 | 121 | ||
AK232201.1 | - | 9 - 10 | 1235 | 121 | ||
AK345180.1 | - | 9 - 10 | 1231 | 121 | ||
AK398895.1 | - | 9 - 10 | 1210 | 121 | ||
U02476.1 | - | 6 - 7 | 899 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |