Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214083.2 | NP_999248.2 | 5 - 6 | 755 | 107 | |
XM_021079496.1 | XP_020935155.1 | 1217 | 107 | |||
XM_021079497.1 | XP_020935156.1 | 1027 | 107 | |||
GenBank mRNA | AB040442.1 | - | 5 - 6 | 755 | 107 | |
AK239062.1 | - | 5 - 6 | 788 | 107 | ||
AK390911.1 | - | 5 - 6 | 792 | 107 | ||
EU026383.1 | - | 5 - 6 | 755 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |