Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001265.1 | NP_001001265.1 | 4 - 5 | 247 | 89 | |
GenBank mRNA | AF148221.1 | - | 4 - 5 | 247 | 89 | |
AK230637.1 | - | 4 - 5 | 276 | 89 | ||
AK230931.1 | - | 4 - 5 | 296 | 89 | ||
AK231001.1 | - | 4 - 5 | 270 | 89 | ||
AK231016.1 | - | 4 - 5 | 296 | 89 | ||
AK231062.1 | - | 4 - 5 | 323 | 89 | ||
AK231092.1 | - | 4 - 5 | 289 | 89 | ||
AK231129.1 | - | 4 - 5 | 270 | 89 | ||
AK343716.1 | - | 4 - 5 | 329 | 89 | ||
AK343723.1 | - | 4 - 5 | 270 | 89 | ||
AK343862.1 | - | 4 - 5 | 270 | 89 | ||
AK343884.1 | - | 4 - 5 | 270 | 89 | ||
AK343896.1 | - | 4 - 5 | 270 | 89 | ||
AK343918.1 | - | 4 - 5 | 289 | 89 | ||
AK343960.1 | - | 4 - 5 | 270 | 89 | ||
AK343964.1 | - | 4 - 5 | 289 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |