Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001628.1 | NP_001001628.1 | 7 - 8 | 921 | 77 | |
XM_005652441.3 | XP_005652498.1 | 2006 | 77 | |||
XM_005652443.3 | XP_005652500.1 | 1835 | 77 | |||
XM_005652444.3 | XP_005652501.1 | 2225 | 77 | |||
XM_005652445.3 | XP_005652502.1 | 1594 | 77 | |||
XM_005652446.3 | XP_005652503.1 | 1263 | 77 | |||
XM_005652447.3 | XP_005652504.1 | 1423 | 77 | |||
XM_013996333.2 | XP_013851787.1 | 2750 | 77 | |||
XM_013996365.2 | XP_013851819.1 | 1060 | 77 | |||
XM_021074699.1 | XP_020930358.1 | 2955 | 77 | |||
XM_021074701.1 | XP_020930360.1 | 366 | 77 | |||
XM_021074702.1 | XP_020930361.1 | 217 | 77 | |||
GenBank mRNA | AB370266.1 | - | 9 - 10 | 1222 | 77 | |
AK238641.1 | - | 7 - 8 | 721 | 77 | ||
AK396797.1 | - | 8 - 9 | 1022 | 77 | ||
AY533021.1 | - | 7 - 8 | 921 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |