Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001631.1 | NP_001001631.1 | 1 - 2 | 78 | 69 | |
GenBank mRNA | AK231951.1 | - | 2 - 3 | 185 | 69 | |
AK239834.1 | - | 2 - 3 | 400 | 69 | ||
AK344909.1 | - | 2 - 3 | 400 | 69 | ||
AK393953.1 | - | 2 - 3 | 192 | 69 | ||
AK399476.1 | - | 2 - 3 | 213 | 69 | ||
AY321067.1 | - | 1 - 2 | 78 | 69 | ||
KU358547.1 | - | 1 - 2 | 56 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |