Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004033.1 | NP_001004033.1 | 2 - 3 | 1000 | 78 | |
XM_005662796.3 | XP_005662853.1 | 1597 | 78 | |||
XM_013996382.2 | XP_013851836.1 | 1341 | 78 | |||
XM_021088739.1 | XP_020944398.1 | 1276 | 78 | |||
XM_021088740.1 | XP_020944399.1 | 1194 | 78 | |||
GenBank mRNA | AK238538.1 | - | 4 - 5 | 1108 | 78 | |
AK351148.1 | - | 4 - 5 | 1108 | 78 | ||
AY134475.1 | - | 2 - 3 | 1000 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Sus scrofa Probe spans exons |