Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005157.1 | NP_001005157.1 | 8 - 9 | 1121 | 64 | |
XM_005665792.3 | XP_005665849.1 | 1357 | 64 | |||
XM_005665793.3 | XP_005665850.1 | 1344 | 64 | |||
XM_005665794.3 | XP_005665851.1 | 1408 | 64 | |||
XM_021098341.1 | XP_020954000.1 | 1788 | 64 | |||
GenBank mRNA | AK236289.1 | - | 9 - 10 | 1295 | 64 | |
AK238677.1 | - | 9 - 10 | 1310 | 64 | ||
AK389641.1 | - | 9 - 10 | 1405 | 64 | ||
AK399598.1 | - | 9 - 10 | 1312 | 64 | ||
EU282340.1 | - | 8 - 9 | 1772 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |