Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011727.1 | NP_001011727.1 | 6 - 7 | 857 | 66 | |
GenBank mRNA | AK234979.1 | - | 6 - 7 | 791 | 66 | |
AK239495.1 | - | 6 - 7 | 891 | 66 | ||
AK346509.1 | - | 1 - 2 | 37 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |