Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031798.1 | NP_001026968.1 | 4 - 5 | 522 | 99 | |
XM_013986257.2 | XP_013841711.1 | 719 | 99 | |||
XM_021079566.1 | XP_020935225.1 | 683 | 99 | |||
GenBank mRNA | AK231821.1 | - | 4 - 5 | 684 | 99 | |
AK238272.1 | - | 4 - 5 | 700 | 99 | ||
AK392729.1 | - | 4 - 5 | 678 | 99 | ||
AY996808.1 | - | 4 - 5 | 522 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |