Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033014.1 | NP_001028186.1 | 1 - 2 | 283 | 99 | |
XM_005653726.3 | XP_005653783.1 | 99 | ||||
GenBank mRNA | AK351654.1 | - | 1 - 2 | 99 | ||
AK394693.1 | - | 1 - 2 | 283 | 99 | ||
DQ003208.1 | - | 1 - 2 | 283 | 99 | ||
KF946098.1 | - | 72 | 99 | |||
KF946099.1 | - | 72 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |