Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037996.1 | NP_001033085.1 | 6 - 7 | 912 | 77 | |
XM_013996822.2 | XP_013852276.1 | 1190 | 77 | |||
GenBank mRNA | AB214979.1 | - | 6 - 7 | 912 | 77 | |
AK232831.1 | - | 6 - 7 | 912 | 77 | ||
AK346033.1 | - | 6 - 7 | 912 | 77 | ||
DQ249306.1 | - | 5 - 6 | 702 | 77 | ||
KF586862.1 | - | 5 - 6 | 696 | 77 | ||
KF586865.1 | - | 5 - 6 | 607 | 77 | ||
KF586866.1 | - | 5 - 6 | 705 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |