Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038006.1 | NP_001033095.1 | 8 - 9 | 1832 | 148 | |
XM_005662947.3 | XP_005663004.1 | 2457 | 148 | |||
XM_013996572.2 | XP_013852026.1 | 2228 | 148 | |||
XM_013996573.2 | XP_013852027.1 | 2316 | 148 | |||
XM_013996574.2 | XP_013852028.1 | 2327 | 148 | |||
GenBank mRNA | DQ091176.1 | - | 8 - 9 | 1832 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |