Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039750.1 | NP_001034839.1 | 2 - 3 | 167 | 99 | |
GenBank mRNA | AB239841.1 | - | 2 - 3 | 178 | 99 | |
AK234275.1 | - | 2 - 3 | 178 | 99 | ||
AK347776.1 | - | 2 - 3 | 154 | 99 | ||
AK392795.1 | - | 2 - 3 | 171 | 99 | ||
DQ402035.1 | - | 2 - 3 | 167 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |