Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UABP-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EW673661.2 | - | 2 - 3 | 197 | 64 | |
X62845.1 | - | 1119 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |