Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213882.1 | NP_999047.1 | 3 - 4 | 468 | 93 | |
GenBank mRNA | AK348522.1 | - | 3 - 4 | 728 | 93 | |
X07383.1 | - | 468 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |