Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213985.1 | NP_999150.1 | 3 - 4 | 1746 | 80 | |
GenBank mRNA | AY038049.1 | - | 3 - 4 | 1746 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |