Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214118.2 | NP_999283.1 | 1 - 2 | 194 | 109 | |
GenBank mRNA | AY116504.1 | - | 1 - 2 | 195 | 109 | |
D21074.1 | - | 1 - 2 | 159 | 109 | ||
DQ108393.1 | - | 1 - 2 | 159 | 109 | ||
U61139.1 | - | 1 - 2 | 159 | 109 | ||
U67175.1 | - | 1 - 2 | 159 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |