Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214125.1 | NP_999290.1 | 2 - 3 | 298 | 110 | |
GenBank mRNA | AF164576.1 | - | 2 - 3 | 298 | 110 | |
KF803270.1 | - | 2 - 3 | 108 | |||
KF803271.1 | - | 2 - 3 | 298 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |