Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214202.1 | NP_999367.1 | 2 - 3 | 101 | 105 | |
XM_005664506.3 | XP_005664563.1 | 177 | 105 | |||
GenBank mRNA | AF152021.1 | - | 2 - 3 | 101 | 105 | |
AK233875.1 | - | 2 - 3 | 124 | 105 | ||
AK344803.1 | - | 2 - 3 | 124 | 105 | ||
AK399683.1 | - | 2 - 3 | 155 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |