Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214238.2 | NP_999403.2 | 2 | 223 | 97 | |
GenBank mRNA | AK349738.1 | - | 2 | 216 | 97 | |
M88166.1 | - | 1 | 151 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |