Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214278.1 | NP_999443.1 | 1 - 2 | 178 | 137 | |
XM_005667890.3 | XP_005667947.1 | 472 | 137 | |||
XM_005667891.3 | XP_005667948.1 | 533 | 137 | |||
XM_005667892.3 | XP_005667949.1 | 543 | 137 | |||
XM_005667893.3 | XP_005667950.1 | 372 | 137 | |||
XM_013979999.2 | XP_013835453.1 | 3437 | 137 | |||
XM_021102240.1 | XP_020957899.1 | 341 | 137 | |||
GenBank mRNA | AF327368.1 | - | 1 - 2 | 178 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |