Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214284.1 | NP_999449.1 | 2 - 3 | 245 | 83 | |
XM_021077291.1 | XP_020932950.1 | 258 | 83 | |||
GenBank mRNA | AF548542.1 | - | 2 - 3 | 245 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |