Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214344.1 | NP_999509.1 | 7 - 8 | 858 | 73 | |
XM_021098311.1 | XP_020953970.1 | 925 | 73 | |||
XM_021098312.1 | XP_020953971.1 | 925 | 73 | |||
XM_021098313.1 | XP_020953972.1 | 889 | 73 | |||
XM_021098314.1 | XP_020953973.1 | 925 | 73 | |||
XM_021098315.1 | XP_020953974.1 | 924 | 73 | |||
GenBank mRNA | AB104465.1 | - | 7 - 8 | 858 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |