Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214346.1 | NP_999511.1 | 15 - 16 | 4023 | 78 | |
XM_021077303.1 | XP_020932962.1 | 4287 | 78 | |||
XM_021077304.1 | XP_020932963.1 | 4289 | 78 | |||
GenBank mRNA | AF509585.1 | - | 15 - 16 | 4023 | 78 | |
DQ176853.1 | - | 15 - 16 | 4023 | 78 | ||
EU003993.1 | - | 15 - 16 | 4023 | 78 | ||
JX070181.1 | - | 15 - 16 | 4023 | 78 | ||
JX070182.1 | - | 15 - 16 | 4023 | 78 | ||
JX070183.1 | - | 15 - 16 | 4023 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |