Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214356.1 | NP_999521.1 | 2 - 3 | 243 | 54 | |
GenBank mRNA | AJ549925.1 | - | 2 - 3 | 243 | 54 | |
AK235024.1 | - | 2 - 3 | 242 | 54 | ||
AY609912.1 | - | 2 - 3 | 194 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |