Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214356.1 | NP_999521.1 | 3 - 4 | 302 | 89 | |
GenBank mRNA | AJ549925.1 | - | 3 - 4 | 302 | 89 | |
AK235024.1 | - | 3 - 4 | 301 | 89 | ||
AY609912.1 | - | 3 - 4 | 253 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |