Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214358.1 | NP_999523.1 | 4 - 5 | 340 | 105 | |
GenBank mRNA | AK230590.1 | - | 3 - 4 | 1918 | 105 | |
AK352180.1 | - | 4 - 5 | 477 | 105 | ||
AK392196.1 | - | 4 - 5 | 528 | 105 | ||
AK399677.1 | - | 4 - 5 | 989 | 105 | ||
AY610311.1 | - | 4 - 5 | 459 | 105 | ||
U57333.1 | - | 4 - 5 | 340 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |