Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214392.2 | NP_999557.2 | 2 - 3 | 364 | 85 | |
GenBank mRNA | AK230563.1 | - | 2 - 3 | 328 | 85 | |
AK230612.1 | - | 2 - 3 | 364 | 85 | ||
AK230919.1 | - | 2 - 3 | 328 | 85 | ||
AK236844.1 | - | 2 - 3 | 333 | 85 | ||
AK343904.1 | - | 2 - 3 | 330 | 85 | ||
EW463673.2 | - | 2 - 3 | 326 | 85 | ||
L14853.1 | - | 2 - 3 | 212 | 85 | ||
U44435.1 | - | 2 - 3 | 340 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |