Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214437.1 | NP_999602.1 | 2 - 3 | 185 | 103 | |
GenBank mRNA | AK238994.1 | - | 2 - 3 | 527 | 103 | |
AY490205.1 | - | 2 - 3 | 185 | 103 | ||
AY509933.1 | - | 2 - 3 | 185 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |