Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213873.2 | NP_999038.2 | 1 - 2 | 72 | 82 | |
GenBank mRNA | EF108301.1 | - | 1 - 2 | 72 | 82 | |
J04601.1 | - | 1 - 2 | 89 | 82 | ||
M20920.1 | - | 1 - 2 | 125 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |