Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130534.1 | NP_001124006.1 | 437 | 79 | ||
GenBank mRNA | AB527060.1 | - | 311 | 79 | ||
AK232197.1 | - | 416 | 79 | |||
AK391026.1 | - | 421 | 79 | |||
AK399232.1 | - | 450 | 79 | |||
EU780066.1 | - | 437 | 79 | |||
FJ439665.1 | - | 312 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa |