Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137631.1 | NP_001131103.1 | 2 - 3 | 942 | 66 | |
XM_021085576.1 | XP_020941235.1 | 1316 | 66 | |||
XM_021085578.1 | XP_020941237.1 | 1234 | 66 | |||
XM_021085579.1 | XP_020941238.1 | 1199 | 66 | |||
XM_021085580.1 | XP_020941239.1 | 1261 | 66 | |||
XM_021085581.1 | XP_020941240.1 | 1326 | 66 | |||
XM_021085582.1 | XP_020941241.1 | 1282 | 66 | |||
XM_021085583.1 | XP_020941242.1 | 1167 | 66 | |||
XM_021085584.1 | XP_020941243.1 | 1169 | 66 | |||
XM_021085585.1 | XP_020941244.1 | 1674 | 66 | |||
XM_021085586.1 | XP_020941245.1 | 1168 | 66 | |||
XM_021085587.1 | XP_020941246.1 | 1167 | 66 | |||
XM_021085589.1 | XP_020941248.1 | 1167 | 66 | |||
XM_021085590.1 | XP_020941249.1 | 1167 | 66 | |||
XM_021085591.1 | XP_020941250.1 | 1166 | 66 | |||
XM_021085592.1 | XP_020941251.1 | 1308 | 66 | |||
XM_021085593.1 | XP_020941252.1 | 1020 | 66 | |||
GenBank mRNA | FJ200453.1 | - | 2 - 3 | 942 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |