Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163801.1 | NP_001157273.1 | 1 - 2 | 512 | 84 | |
GenBank mRNA | AK351135.1 | - | 1 - 2 | 515 | 84 | |
AK351334.1 | - | 1 - 2 | 516 | 84 | ||
AK393112.1 | - | 1 - 2 | 510 | 84 | ||
AK398310.1 | - | 1 - 2 | 510 | 84 | ||
GQ266680.1 | - | 1 - 2 | 512 | 84 | ||
KX779146.1 | - | 2 - 3 | 221 | 84 | ||
KX839662.1 | - | 2 - 3 | 221 | 84 | ||
KX839663.1 | - | 2 - 3 | 347 | 84 | ||
KX839664.1 | - | 2 - 3 | 416 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |