Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166490.1 | NP_001159962.1 | 4 - 5 | 488 | 79 | |
GenBank mRNA | GQ996936.1 | - | 4 - 5 | 488 | 79 |
Gene Symbol: LOC110255217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097830.1 | XP_020953489.1 | 488 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |