Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167638.1 | NP_001161110.1 | 6 - 7 | 755 | 84 | |
GenBank mRNA | AB527054.1 | - | 6 - 7 | 755 | 84 | |
AK232273.1 | - | 6 - 7 | 783 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |