Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128472.1 | NP_001121944.1 | 3 - 4 | 445 | 73 | |
GenBank mRNA | AK347456.1 | - | 3 - 4 | 478 | 73 | |
AK347693.1 | - | 3 - 4 | 474 | 73 | ||
EF581989.1 | - | 3 - 4 | 445 | 73 | ||
EU429459.1 | - | 3 - 4 | 1373 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |