Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128473.1 | NP_001121945.1 | 4 - 5 | 589 | 95 | |
XM_021088503.1 | XP_020944162.1 | 621 | 95 | |||
XM_021088504.1 | XP_020944163.1 | 608 | 95 | |||
GenBank mRNA | AK239047.1 | - | 4 - 5 | 511 | 95 | |
AK391444.1 | - | 4 - 5 | 564 | 95 | ||
EU259887.1 | - | 4 - 5 | 589 | 95 | ||
KF977840.1 | - | 3 - 4 | 420 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |