Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128473.1 | NP_001121945.1 | 5 - 6 | 787 | 87 | |
XM_021088503.1 | XP_020944162.1 | 819 | 87 | |||
XM_021088504.1 | XP_020944163.1 | 806 | 87 | |||
GenBank mRNA | AK234627.1 | - | 5 - 6 | 791 | 87 | |
AK239047.1 | - | 5 - 6 | 709 | 87 | ||
AK391444.1 | - | 5 - 6 | 762 | 87 | ||
AK393077.1 | - | 1 - 2 | 613 | 87 | ||
EU259887.1 | - | 5 - 6 | 787 | 87 | ||
KF977840.1 | - | 4 - 5 | 618 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |