Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142841.1 | NP_001136313.1 | 2 - 3 | 139 | 104 | |
GenBank mRNA | FJ424703.1 | - | 2 - 3 | 139 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |