Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143703.1 | NP_001137175.1 | 4 - 5 | 483 | 87 | |
GenBank mRNA | FJ436394.1 | - | 4 - 5 | 483 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |