Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143723.1 | NP_001137195.1 | 1 - 2 | 92 | 64 | |
GenBank mRNA | FJ436381.1 | - | 1 - 2 | 92 | 64 | |
FJ436382.1 | - | 1 - 2 | 87 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |