Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144842.2 | NP_001138314.1 | 4 - 5 | 472 | 91 | |
GenBank mRNA | AK391822.1 | - | 4 - 5 | 500 | 91 | |
FJ487633.1 | - | 4 - 5 | 473 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |