Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159592.1 | NP_001153064.1 | 2 - 3 | 340 | 75 | |
GenBank mRNA | AK352367.1 | - | 2 - 3 | 359 | 75 | |
AY609739.1 | - | 2 - 3 | 340 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |