Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160081.1 | NP_001153553.1 | 2 - 3 | 528 | 82 | |
XM_021068338.1 | XP_020923997.1 | 441 | 82 | |||
GenBank mRNA | FJ875103.1 | - | 2 - 3 | 528 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |