Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160087.1 | NP_001153559.1 | 5 - 6 | 1060 | 96 | |
XM_021098154.1 | XP_020953813.1 | 1190 | 96 | |||
XM_021098155.1 | XP_020953814.1 | 1490 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |