Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161648.1 | NP_001155120.1 | 4 - 5 | 274 | 59 | |
GenBank mRNA | AK237722.1 | - | 4 - 5 | 364 | 59 | |
FJ968780.1 | - | 4 - 5 | 274 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |