Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172363.2 | NP_001165834.1 | 2 - 3 | 240 | 77 | |
XM_013994501.2 | XP_013849955.2 | 890 | 77 | |||
XM_021081651.1 | XP_020937310.1 | 323 | 77 | |||
XM_021081655.1 | XP_020937314.1 | 631 | 77 | |||
XM_021081659.1 | XP_020937318.1 | 452 | 77 | |||
GenBank mRNA | AK236433.1 | - | 2 - 3 | 278 | 77 | |
AK343612.1 | - | 2 - 3 | 231 | 77 | ||
AK343811.1 | - | 2 - 3 | 229 | 77 | ||
AK391205.1 | - | 2 - 3 | 231 | 77 | ||
AK391208.1 | - | 2 - 3 | 228 | 77 | ||
AK391657.1 | - | 2 - 3 | 232 | 77 | ||
AK395255.1 | - | 2 - 3 | 264 | 77 | ||
AK399432.1 | - | 2 - 3 | 232 | 77 | ||
AK399447.1 | - | 2 - 3 | 264 | 77 | ||
AK399466.1 | - | 2 - 3 | 231 | 77 | ||
AK399472.1 | - | 2 - 3 | 247 | 77 | ||
AK399544.1 | - | 2 - 3 | 231 | 77 | ||
AK399555.1 | - | 2 - 3 | 231 | 77 | ||
AK399560.1 | - | 4 - 5 | 440 | 77 | ||
FJ865398.1 | - | 2 - 3 | 241 | 77 | ||
U07178.1 | - | 2 - 3 | 227 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |